Learn More
TARGETMOL CHEMICALS INC MEAIB 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T4019450MG
Encompass_Preferred
Also available in 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MeAIB is an N-acetyltransferase inhibitor that inhibits SNAT2-related signaling pathways. MeAIB is a specific substrate for amino acid transport system a (ATA1). Amino acid transport system a (ATA1) regulates the uptake of short chain neutral amino acids by Na+ flow. purity: 99%
Catalog No. 50-258-0000