Learn More
MSC Cortech USA Meal Tray Delivery Cart, Insulated, Stainless Steel Base
Supplier:  MSC 38196168
Encompass
This one-piece, insulated meal tray delivery cart is designed for high impact resistance and frequent use. Its walls and doors provide maximum thermal retention when filled with various tray types. The solid one-piece sliding doors are easily removable for cleaning and facilitate safe and efficient meal delivery.
  • Accommodates trays up to 9 1/2 in x 14 1/2 in.
  • Raised top edge surface for safely holding more trays, beverage servers, and wash racks.
  • Push/pull handles for easy navigation.
  • Optional tamperproof locking door latch system.
  • Highest quality non-marring kingpin-less casters.
  • Holds up to 150 meal trays (84 with lid).
  • Built-in wall bumpers.
  • Solid one-piece sliding doors.
  • No dangerous hinges.
  • Stainless steel base.
  • Manufactured to ISO quality standards in the USA.
Catalog No. 50-283-8191