Learn More
Grainger Measure Miser 12 3/4 in L Solid PK12
Supplier:  Grainger 14D251
Encompass_Preferred
The Carlisle Measure Miser is a 3 ounce capacity plastic measuring cup with clear graduations up to 3 ounces in 1/8 ounce increments. It is made of durable polypropylene plastic and has a pour spout with an attached cap. The cup has a wide base for stability and is top-rack dishwasher safe. It can be used to accurately measure small amounts of liquid ingredients for cooking, baking, arts and crafts, and other applications requiring precise measurement up to 3 fluid ounces.
Catalog No. 50-275-2481