Learn More
Electron Microscopy Sciences Measuring Scale #8, for 15X Measuring Magnifier, 26 mm
SDP
Supplier:  Electron Microscopy Sciences 6817515X
Encompass_Preferred
These scales are for use with the Measuring Magnifiers. Measuring scale for 15x magnifiers. 0 to 20mm (0.1mm divisions), 15X Scale: 0 to 14mm (0.1mm divisions)
Use: 15X
Size: 26 mm
Catalog No. 50-192-8515