Learn More
Medchemexpress LLC Mebeverine acid-d5 | 00-00-0 | 98.0% | 284.41 g/mol | C16H20D5NO3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY12769S15MG
Encompass_Preferred
Mebeverine acid-d5 is the deuterium-labeled metabolite of mebeverine supplied as an analytical reference standard for tracer, pharmacokinetic, and metabolism studies. The labeled analogue provides a predictable mass shift for use in LC-MS/MS assays, method development, and quantitative analysis when used alongside a certificate of analysis.
  • Deuterium-labeled metabolite for tracer and quantitation studies.
  • Suitable for LC-MS/MS and pharmacokinetic method development.
  • High isotopic and chemical purity provided with a certificate of analysis.
  • Stable under recommended storage; follow COA and SDS for handling.
  • Available in small milligram quantities for analytical workflows.
Catalog No. 50-002-64385