Learn More
Medchemexpress LLC Mebeverine alcohol | 14367-47-6 | 99.6% | 1 ML

Supplier: Medchemexpress LLC HY1277010MM/1ML
Mebeverine alcohol is a metabolite of Mebeverine, a musculotropic antispasmodic drug. This chemical is provided as a 10 mM solution in 1 mL of DMSO, offering a high purity of 99.6%.
- Solution provided at 10 mM in 1 mL DMSO
- High purity of 99.6%
- Appears as a light yellow to yellow liquid
- Soluble in DMSO at 100 mg/mL
- Stable under recommended storage conditions
- Not classified as a hazardous substance or mixture
- Protocols for in vivo dissolution are available
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.