Learn More
Medchemexpress LLC Mebeverine alcohol | 14367-47-6 | 99.6% | 100 MG

Supplier: Medchemexpress LLC HY12770100MG
Mebeverine alcohol is a metabolite of Mebeverine, a musculotropic antispasmodic drug. This product appears as a liquid, ranging from light yellow to yellow. For optimal stability, the pure form should be stored at -20°C for up to 3 years or 4°C for up to 2 years. If in solvent, store at -80°C for 6 months or -20°C for 1 month.
- High purity of 99.6%
- Molecular formula: C16H27NO2
- Molecular weight: 265.39
- CAS number: 14367-47-6
- Appears as a light yellow to yellow liquid
- Soluble in DMSO at 100 mg/mL
- Classified as a drug metabolite and metabolic enzyme/protease
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.