Learn More
Medchemexpress LLC Mebeverine alcohol | 14367-47-6 | 99.6% | 50 MG

Supplier: Medchemexpress LLC HY1277050MG
Mebeverine alcohol is a metabolite of Mebeverine, a musculotropic antispasmodic drug. It has a molecular weight of 265.39 and a chemical formula of C16H27NO2. This product appears as a liquid, specifically light yellow to yellow in color, and is intended for research use only.
- Metabolite of Mebeverine, a musculotropic antispasmodic drug.
- Chemical formula: C16H27NO2.
- Molecular weight: 265.39.
- Appearance: Liquid.
- Color: Light yellow to yellow.
- High purity: 99.61%.
- Solubility: ≥ 2.5 mg/mL (9.42 mM) in various in vivo dissolution protocols.
- Storage conditions for pure form: -20°C for 3 years, 4°C for 2 years.
- Storage conditions in solvent: -80°C for 6 months, -20°C for 1 month.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.