Learn More
Medchemexpress LLC Mebeverine hydrochloride | 2753-45-9 | 99.85% | C25H36ClNO5 | 100 MG

Supplier: Medchemexpress LLC HYA0078100MG
Mebeverine hydrochloride is a β-phenylethylamine derivative and a musculotropic agent. It potently blocks intestinal peristalsis by directly blocking voltage-operated sodium channels and inhibiting intracellular calcium accumulation.
- Molecular weight: 466.01
- Appearance: Solid
- Color: White to off-white
- Soluble in DMSO at ≥ 155 mg/mL (332.61 mM)
- Inhibits alpha1 action by limiting calcium mobilization on receptor stimulation in guinea-pig taenia caeci smooth muscle cells
- Exhibits atropine-like properties at 6x10^-6 M
- Causes a small hyperpolarization of muscle cells (2.1 + 0.5 mV) and suppresses spike activity at 6x10^-5 M
- Related to metabolic or endocrine disease, endocrinology
- Acts on sodium channel
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.