Learn More
Grainger Mechanical Food Service Thermometer
Supplier:  Grainger 14F307
Encompass_Preferred
The Rubbermaid Commercial Products Mechanical Food Service Thermometer is a stainless steel thermometer with a temperature range of 0 to 220 degrees Fahrenheit. It has a 5 inch stem and a 1.5 inch dial display. The thermometer is NSF certified and features a pointed, stepped probe tip for easy insertion. It can be calibrated and is dishwasher safe. This thermometer allows quick and accurate temperature readings for commercial kitchen use.
Catalog No. 50-275-0821