Learn More
Grainger Mechanics Gloves Black/Gray 10 PR
Supplier:  Grainger 16V389
Encompass_Preferred
The Mechanix Wear M-Pact Covert Tactical Gloves are a pair of durable work gloves designed for mechanics, industrial workers, and tactical operators. They feature a seamless single layer Clarino synthetic leather palm that is abrasion resistant. The back of the hand uses four-way stretch Spandex material for flexibility. The gloves have molded rubberized knuckle protection and finger sidewalls for impact resistance. They also have a Thermal Plastic Rubber (TPR) closure for a secure fit and hook/loop wrist closure. The M-Pact Covert Tactical Gloves come in sizes S-2XL in black and coyote colors.
Catalog No. 50-276-6713