Learn More
Seca Corp 222 Mechanical telescopic measuring rod with large measuring range, cm-only
Supplier: Seca Corp 222171408
Specially developed for wall mounting, this measuring rod is aligned to work exactly to the millimeter from head to toe. Its broad measuring slide and heel positioner are professional prerequisites. The telescopic measuring rod ensures that the result can be easily read at eye level even for the tallest patient. When not in use, the measuring slide can be folded down for safety. • Telescopic, durable aluminum measuring rod with large measurement range up to 90 inches • Includes heel positioner for ideal three-point measurement • Developed for wall installation • Measuring range: 6 – 230 cm • Graduation: 1 mm
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.