Learn More
Medchemexpress LLC Mecillinam (amdinocillin) | 32887-01-7 | 95.0% | C15H23N3O3S | 100 MG

Supplier: Medchemexpress LLC HYA0269100MG
Mecillinam (Amdinocillin) is a β-lactam antibiotic that demonstrates broad-spectrum activity against gram-negative organisms. It is intended for research use only.
- Broad-spectrum activity against gram-negative organisms
- Solid appearance
- White to off-white color
- Soluble in DMSO and H2O
- Storage at 4°C, protected from light, under nitrogen
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.