Learn More
ABclonal Technology Meckelin Rabbit mAb
SDP
Supplier:  ABclonal Technology A21162100UL
Encompass_Preferred
The protein encoded by this gene localizes to the primary cilium and to the plasma membrane The gene functions in centriole migration to the apical membrane and formation of the primary cilium Multiple transcript variants encoding different isoforms have been found for this gene Defects in this gene are a cause of Meckel syndrome type 3 (MKS3) and Joubert syndrome type 6 (JBTS6)
Catalog No. 50-000-07585