Learn More
Cayman Chemical MeclIzIn N-oxIde 5mg
SDP
Supplier:  Cayman Chemical 350545 MG
Encompass_Preferred
A metabolite of meclizine; a degradation product and potential impurity in commercial preparations of meclizine
Catalog No. 50-291-0966