Learn More
TARGETMOL CHEMICALS INC Meclizine 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T406521MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Meclizine, also known as Meclozine, is a piperazine class H1 antagonist with antihistamine properties that inhibits the interaction of histamine at H1 receptors reversibly. It serves as an effective anti-motion sickness agent, able to cross the blood-brain barrier. Additionally, Meclizine acts as an agonist ligand for the mouse constitutive androstane receptor (CAR) and an inverse agonist for human CAR, making it useful in research on polyQ toxicity disorders, including Huntington's disease. Purity 95.16%
Catalog No. 50-288-0454