Learn More
Selleck Chemical LLC Meclofenoxate (Centrophenoxine) HCl S4280-200mg
SDP
Supplier:  Selleck Chemical LLC S4280200MG
Encompass_Preferred
Meclofenoxate (Centrophenoxine) HCl is an anti-aging drug used to treat the symptoms of senile dementia and Alzheimer s disease and also inhibits the activity of cholinephosphotransferase
Catalog No. 50-310-9428