Learn More
Medchemexpress LLC MeCY5-NHS ester triethylamine | 00-00-0 | 95.3% | C42H56N4O10S2 | 10MG
SDP
Supplier:  Medchemexpress LLC HY135413B10MG
Encompass_Preferred
The triethylamine salt of a Cy5-family N-hydroxysuccinimide (NHS) ester for covalent fluorescent labeling of biomolecules. It reacts with primary amines on proteins, peptides, and oligonucleotides to form stable amide linkages, enabling far-red fluorescence detection in imaging and biochemical assays. The reagent is supplied in research-scale vials for conjugation workflows.
  • Reacts with primary amines to form stable amide bonds.
  • Suitable for labeling proteins, peptides, and oligonucleotides.
  • Cy5-family fluorophore for far-red detection.
  • Supplied as a triethylamine salt for handling and solubility.
  • Available in small pack sizes for research use.
  • Purity 95.3%.
Catalog No. 50-002-59967