Learn More
Apexbio Technology LLC Medetomidine HCl 86347-15-1 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B13615.1
Encompass_Preferred
Medetomidine HCl (CAS 86347-15-1) is a potent and highly selective agonist of the 2-adrenoceptor with a binding affinity (K sub i /sub ) of 1 08 nM Demonstrating approximately 1620-fold greater selectivity for 2- over 1-adrenoceptors medetomidine modulates adrenergic signaling predominantly via presynaptic inhibition of norepinephrine release This selectivity makes medetomidine a valuable tool for studies involving 2-adrenergic receptor pharmacology including research on neurotransmitter modulation central nervous system function and cardiovascular physiology
Catalog No. 50-000-92518