Learn More
Cayman Chemical MedetomIdInhydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 1645410 MG
Encompass_Preferred
An α2-AR agonist (Ki = 1.08 nM); selective for α2-ARs over α1-ARs (Ki = 1,750 nM); induces sedation in dogs
Catalog No. 50-297-1929