Learn More
Cliawaived Inc N3100 Series Nitrile Exam Gloves LARGE. Powder free, exam grade, Chemo tested (1 box of 100 gloves)
SDP
Supplier:  Cliawaived Inc VAL3100L
Encompass_Preferred
N3100 Series Nitrile Exam Gloves Large. Powder free, exam grade, Chemo tested. Dependable for house-wide applications within Healthcare, Labs, EMS and other settings where high-use and disposable exam glove protection is important. Size: Extra LargeNitrileThickness (Finger): 3.5milPowder FreeASTM D6319, D6978Exam GradeColor: BlueTexture: FingertipLength: Standard (240mm)Cuff: BeadedNon-SterileChemotherapy Tested**Chemotherapy drug testing information on back of box(1 box of 100)
Catalog No. NC2580177