Learn More
TARGETMOL CHEMICALS INC MEDIFOXAMINE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T244445MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Medifoxamine (LG 152) is a selective non-monoamine oxidase inhibitor that exhibits antidepressant activity through inhibition of 5 HT reuptake.Medifoxamine preferentially inhibits dopamine reuptake. purity: 98%
Catalog No. 50-258-0807