Learn More
Electron Microscopy Sciences Medium Handle Needle File
SDP
Supplier:  Electron Microscopy Sciences 6221302
Encompass_Preferred
Blue anodized aluminum handles are well knurled with an excellent grip. The length of the handle is 4" and features a steel easy-release chuck, which simply presses into the barrel and remains secure. Handle will last a lifetime Made in the USA. Length: 5-1/2 - 6-1/4", diameter: 1/2".
Catalog No. 50-306-48