Learn More
Enterprise Technology Solutions Medline Face Mask Shield,Fluid-Resistant,Nonglare,Fog-Free,25/BX,PE
SDP
Supplier:  Enterprise Technology Solutions MIINON27410EL
Encompass_Preferred
Surgical masks feature a protective eyeshield to provide full protection against fluids at either the ASTM F1862 160 mmHg or 120 mmHg fluid resistance levels. Attached eyeshield provides protection over eyes or glasses. Nonglare, wraparound eyeshield assures maximum eye and facial protection. The polypropylene outer facing provides added protection against fluids while the inner cellulose facing adds comfort. Anti-fog strip is made of comfortable, soft foam to prevent eyeglasses from fogging, ensuring optical clarity, while the ties provide a secure attachment.
Catalog No. 50-008-28102