Learn More
Enterprise Technology Solutions Medline Safety Glasses,UV/Impact Protection,CL Frame/Lens
SDP
Supplier:  Enterprise Technology Solutions MIINON24770
Encompass_Preferred
Clear Frame/Lens Safety Glasses provide optical clarity. They are ANSI Z87.1 impact-rated to protect your eyes from impact and are ANSI Z87.1 U6-rated, providing 99.9 percent UV filtration. Safety glasses fit comfortably and securely to your face.
Catalog No. 50-008-27898