Learn More
Medchemexpress LLC Medrate-d3 | 1262969-27-6 | 377.49 g/mol | C22H27D3O5 | 1 MG

Supplier: Medchemexpress LLC HYW6542671MG
Medrate-d3 is the deuterium-labeled (stable isotope) form of Medrate supplied for laboratory research as an isotope-labeled reference or tracer. Intended for analytical and isotopic-labeling applications; not for human use. Lot-specific purity and certificate of analysis (COA) are provided by the manufacturer.
- Deuterium-labeled reference compound for analytical and tracer applications
- Chemical formula C22H27D3O5
- Molecular weight 377.49 g/mol
- CAS number 1262969-27-6
- Provided in small quantities suitable for analytical standards (e.g., 1 mg)
- Purity and storage conditions reported on lot-specific certificate of analysis
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.