Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Medroxyprogesterone acetate | 71-58-9 | 99.9% | 100 MG
SDP

Supplier:  Medchemexpress LLC HYB0469100MG

Encompass_Preferred

Medroxyprogesterone acetate is a synthetic steroid widely used due to its interaction with progesterone, androgen, and glucocorticoid receptors. It is a white to off-white solid with a molecular formula of C24H34O4 and a molecular weight of 386.52.

  • Interacts with progesterone, androgen, and glucocorticoid receptors
  • Molecular formula C24H34O4
  • Molecular weight 386.52
  • White to off-white solid appearance
  • Soluble in DMSO and ethanol
  • Inhibits eNOS expression in HUVEC cells
  • Reduces NF-κB nuclear translocation
  • Exhibits antiproliferative activity in multiple human cell lines
  • Agonist for human progesterone, glucocorticoid, and androgen receptors

Catalog No. 50-004-08220


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.