Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Medroxyprogesterone acetate (Standard) | 71-58-9 | 99.2% | 25 MG
SDP

Supplier:  Medchemexpress LLC 25MGHYB0469R

Encompass_Preferred

Medroxyprogesterone acetate (Standard) is the analytical standard of Medroxyprogesterone acetate, a widely used synthetic steroid that interacts with progesterone, androgen, and glucocorticoid receptors. This compound is intended for research and analytical applications, commonly utilized in qualitative, quantitative, and methodological research experiments, including HPLC, GC, and MS, as a reference standard.

  • Molecular weight: 386.52
  • Formula: C₂₄H₃₄O₄
  • Appearance: Solid
  • Color: White to off-white
  • Structure classification: Steroids
  • Initial source: Endogenous metabolite
  • Application: Analytical standard for assay

Catalog No. 50-004-50587


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.