Learn More
Medchemexpress LLC Mefenamic acid | 61-68-7 | 99.9% | 5 G

Supplier: Medchemexpress LLC HYB05745G
Mefenamic acid is a non-steroidal anti-inflammatory agent that is permeable to the blood-brain barrier. It acts as a competitive inhibitor of human cyclooxygenase-1 (hCOX-1) and human cyclooxygenase-2 (hCOX-2).
- Acts as a competitive inhibitor of hCOX-1 and hCOX-2.
- Has cytotoxic effects on KB, Saos-2, and 1321N cells.
- Molecular weight: 241.29.
- Chemical formula: C15H15NO2.
- Appears as a white to off-white solid.
- Soluble in DMSO at ≥ 100 mg/mL.
- Insoluble in water.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.