Learn More
Medchemexpress LLC Mefluleucine | 25MG
SDP
Supplier:  Medchemexpress LLC HY11438425MG
Encompass_Preferred
Mefluleucine (NV-5138) a leucine analog is the first selective and orally active brain mTORC1 activator binding to Sestrin2 Mefluleucine is used for antidepressant studies[1][2]
Catalog No. 50-002-28009