Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mefluleucine hydroch 10mM 1mL | 2639392-70-2 | 1 ML
SDP

Supplier:  Medchemexpress LLC HY114384B10MM 1ML

Encompass_Preferred

Mefluleucine hydrochloride is a leucine analog that selectively activates brain mTORC1 by binding Sestrin2. It is used in antidepressant and neuroscience research.

  • Leucine analog and selective brain mTORC1 activator.
  • Used for antidepressant and neuroscience research.
  • Purity typically 98.0% (NMR).
  • Molecular weight ≈ 217.64 g/mol; formula C7H14ClF2NO2.
  • Soluble in DMSO (≈100 mg/mL) and compatible with common in vivo formulations at ≥2.08 mg/mL (≈9.56 mM).
  • Store solid at 4°C, sealed and protected from moisture; in solution store at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-28861


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.