Learn More
Medchemexpress LLC Mefuparib | 1392502-82-7 | 99.8% | 298.31 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY167927
Encompass_Preferred
Mefuparib (CVL218) is a poly ADP-ribose polymerase (PARP) inhibitor used in cancer research that exhibits potent brain penetration due to its high protein binding.
  • Acts as a poly ADP-ribose polymerase (PARP) inhibitor.
  • Used in cancer research.
  • Exhibits potent brain penetration.
  • Known for its high protein binding.
Catalog No. 50-003-94685