Learn More
AdipoGen MEGA-8 ultrapure
SDP
Supplier:  AdipoGen AGCC10007G005
Encompass_Preferred
Chemical. CAS 85316-98-9. Formula C15H31NO6. MW 321.4. Non-ionic water-soluble detergent with nondenaturing properties. Transparent in the UV region. Relatively high critical micelle concentration CMC value 79mM/l. Ideal for studying the conformation and function of proteins, keeping the secondary and tertiary structure intact, therefore suited for solubilizing membrane proteins without altering biological activity. Efficiently separates hydrophilic proteins from membrane spanning, hydrophobic proteins. Ideal for use as membrane protein solubilizer.
Catalog No. 50-256-2711