Learn More
Leica Biosystems MEGACASSETTES WHITE 1000
Supplier:  Leica Biosystems 38VSP59040
Encompass
These cassettes offer double the depth of conventional cassettes.Allows for easier embedding and processing of larger or whole organs (such as eyes, teeth and bones), as well as ossified tissue that do not fit standard cassettes.Perfect for research and specialty clinical applications.The cassette dimensions are 40 x 26 x 13 mm and are available in 6 colors in quantities of 1,000.Can be used with the Mega cassettes base mold. Made of stainless steel, these reusable base molds allow easy block removal after cooling. Available in boxes of 10.
Catalog No. NC1882916