Learn More
Medchemexpress LLC Meglumine | 6284-40-8 | 98.0% | 10 G
SDP
Supplier:  Medchemexpress LLC HYB034210G
Encompass_Preferred
Meglumine (Methylglucamine) is an orally active amino sugar derived from sorbitol, exhibiting anti-inflammatory and antitumor activity. It is frequently used as an excipient in active molecules and with iodinated compounds in contrast agents. This product is for research use only.
  • Reduces inflammatory factors in THP-1 human myeloid cells and RAW264.7 mouse macrophage cells
  • Increases SNARK expression levels in C2C12 mouse myoblasts
  • Reduces inflammatory factors and inhibits skin cancer tumor growth in rats and mice
  • Improves muscle function, limits metabolic syndrome, and reduces diabetic complications in type 2 diabetic mice
Catalog No. 50-004-06213