Learn More
Medchemexpress LLC Meglumine (Methylglucamine) | 6284-40-8 | 98.0% | 5 G
SDP
Supplier:  Medchemexpress LLC HYB03425G
Encompass_Preferred
Meglumine (Methylglucamine) is an orally active amino sugar derived from sorbitol. It exhibits anti-inflammatory and antitumor activities, making it suitable for various research applications. It is frequently utilized as an excipient in active molecules and in conjunction with iodinated compounds in contrast agents, such as meglumine and meglumine iodide.
  • Derived from sorbitol
  • Exhibits anti-inflammatory activity
  • Exhibits antitumor activity
  • Utilized as an excipient in active molecules
  • Used in contrast agents with iodinated compounds
Catalog No. 50-004-07254