Learn More
Perkin Elmer US LLC MEINHARD Plus Nebulizer Internal Standard Tee
Supplier:  Perkin Elmer US LLC N8145510
Encompass_Preferred
The internal standard mixing tee is perfect for online addition of internal standard or online dilution. Achieves excellent mixing with minimal dead volume.
Catalog No. 50-211-1901