Learn More
Medchemexpress LLC HY-134259 10mg , MEISi-2 CAS:2250156-71-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13425910MG
Encompass_Preferred
HY-134259 10mg MEISi-2 CAS: 2250156-71-7 MEISi-2 is a selective inhibitor of MEIS, a key regulator of hematopoietic stem cell (HSC) self-renewal. MEISi-2 is developed for the research of cardiac injuries, hematopoiesis issues, bone marrow transplantations, and cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0061