Learn More
ABclonal Technology MEK1/MEK2 Rabbit mAb
SDP
Supplier:  ABclonal Technology A2439420UL
Encompass_Preferred
The protein encoded by this gene is a member of the dual specificity protein kinase family which acts as a mitogen-activated protein (MAP) kinase kinase MAP kinases also known as extracellular signal-regulated kinases (ERKs) act as an integration point for multiple biochemical signals This protein kinase lies upstream of MAP kinases and stimulates the enzymatic activity of MAP kinases upon wide variety of extra- and intracellular signals As an essential component of MAP kinase signal transduction pathway this kinase is involved in many cellular processes such as proliferation differentiation transcription regulation and development [provided by RefSeq Jul 2008]
Catalog No. 50-000-09390