Learn More
Medchemexpress LLC HY-15202 200mg , MEK162 Binimetinib;ARRY-162;ARRY-438162 CAS:606143-89-9 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY15202200MG
Encompass_Preferred
Medchemexpress, HY-15202 200mg MEK162 Binimetinib;ARRY-162;ARRY-438162 CAS:606143-89-9 Formula:C17H15BrF2N4O3 IC50: 12 nM (MEK) Purity:98% MEK162 is a potent and selective mitogen-activated protein kinase ( MEK ) inhibitor wirh IC 50 of 12 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2564