Learn More
Enzo Life Sciences MEKK 1 polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS2930100
Encompass_Preferred
Immunogen: Recombinant MEKK1 (aa 1307-1509). Species reactivity: Human. Applications: ELISA, ICC, IHC (FS), IHC (PS), WB. Purity: Affinity purified. Formulation: Liquid. In PBS, pH 7.4, containing 0.02% sodium azide and 50% glycerol.
Catalog No. 50-201-2833