Learn More
Medchemexpress LLC MEL24 | 642072-48-8 | C17H18N4O3 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY15321510MM/1ML
Encompass_Preferred
MEL24 is an Mdm2 E3 ligase inhibitor that reduces cell survival and increases sensitivity to DNA damaging agents in a p53-dependent manner for in vitro antitumor studies. It is for research use only.
  • Mdm2 E3 ligase inhibitor
  • Reduces cell survival
  • Increases sensitivity to DNA damaging agents
  • P53-dependent action
  • For in vitro antitumor studies
  • Acts on 293T cells with ED50 value of 9.2 μM
  • Activates p53 in HCT116 cells
Catalog No. 50-003-72613