Learn More
Medchemexpress LLC Melamine | 108-78-1 | MFCD00006055 | 100.0% | 126.12 g/mol | C3H6N6 | 100g
SDP
Supplier:  Medchemexpress LLC HYY1117100G
Encompass_Preferred
Melamine is an orally active inducer of Apoptosis Melamine induces animal disease models Melamine affects the activity of Sertoli cell and can be used for research on male reproductive function Melamine also has neurotoxicity and nephrotoxicity Melamine induces cognitive impairment and acute kidney injury models Melamine can also be used to induce bladder cancer and urinary stone models[1][2][3][4][5][6]
Catalog No. 50-002-68117