Learn More
Medchemexpress LLC Melamine (13C3,15N3) | 1246816-14-7 | 96.1% | 132.08 | 13C3H6N315N3 | 1mg
SDP
Supplier:  Medchemexpress LLC HYY1117S21MG
Encompass_Preferred
Melamine-15N3 13C3 is a 13C- and 15N-labeled Melamine (HY-Y1117) Melamine is a metabolite of cyromazine Melamine is a intermediate for the synthesis of melamine resin and plastic materials[1][2]
Catalog No. 50-002-67130