Learn More
U.S. Pharmacopeia Melamine, 108-78-1, MFCD00006055, 250mg
Supplier:  U.S. Pharmacopeia 1379183
Encompass
Molecular formula C3H6N6, Molecular weight 126.13 g/mol, Melting Point 669.2 °F (354 °C) (decomposes and sublimes at about 280 ° C). Synonyms: Cyanurotriamide
Catalog No. NC0281335