Learn More
Genetex Inc Melan A Monoclonal Antibody [MSVA-901M+] HistoMAX™ (GTX04384) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04384
Encompass_Preferred
The Melan A antibody [MSVA-901M+] HistoMAX™ (GTX04384) is a Mouse Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human malignant melanoma from skin tissue using GTX04384 Melan A antibody [MSVA-901M+] HistoMAX. Malignant melanoma with strong Melan A positivity in all tumor cells.
Catalog No. NC3666313