Learn More
Genetex Inc Melanoma gp100 Monoclonal Antibody [MSVA-617R] HistoMAX™ (GTX04382) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04382
Encompass_Preferred
The Melanoma gp100 antibody [MSVA-617R] HistoMAX™ (GTX04382) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human skin tissue using GTX04382 Melanoma gp100 antibody [MSVA-617R] HistoMAX. Strong melanoma gp100 immunostaining in melanocytes of the skin.
Catalog No. NC3666304