Learn More
BIO SB INC Melanoma Cocktail (HMB-45, A103 & BSB-6), MMab
Supplier:  BIO SB INC BSB 6882
Encompass
Melanoma Cocktail is a mouse monoclonal antibody cocktail derived from cell culture supernatant that is concentrated, dialyzed, filter sterilized and diluted in buffer pH 7.5, containing BSA and sodium azide as a preservative.
Catalog No. NC2828682