Learn More
Discovery Life Sciences Melanoma DTC
SDP
Supplier:  Discovery Life Sciences 320410
Encompass
DTC Dissociated Tumor and Tissue Cells start as fresh solid tumor tissue collected during surgery are enzymatically and mechanically dissociated into single cells and are delivered as viably cryopreserved cells for functional assays cell culture and single cell genomics
Catalog No. 50-008-04055