Learn More
Medchemexpress LLC Melarsoprol | 494-79-1 | 98.0% | 1 MG

Supplier: Medchemexpress LLC HY1195941MG
Melarsoprol is a trivalent organic arsenical research reagent used primarily as an antitrypanosomal agent; it can cross the blood-brain barrier and induces apoptosis in certain leukemic cells. Supplied as a powdered research-grade compound for laboratory use.
- Antitrypanosomal activity; crosses the blood-brain barrier.
- Research-grade powder, purity 98.0%.
- Molecular formula C12H15AsN6OS2; molecular weight 398.34 g/mol.
- Physical form: powder; recommended storage: -20°C (powder, stable 3 years) or 4°C (powder, stable 2 years).
- Intended for research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.